Cerium(IV) oxide ≥99.995% (Ce basis), PURATREM

Supplier: STREM CHEMICALS INC MS

Synonyms: Ceric oxide, Cerium dioxide

58-0800-25G 58-0800-5G
100210-026EA 213.72 USD
100210-026 100196-316
Cerium(IV) oxide ≥99.995% (Ce basis), PURATREM
Cerium(IV) oxide
Formula: CeO₂
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: ∼2600 °C
Density: 7.13 g/cm³ (25 °C)
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR