Cerium tetrafluoride, anhydrous ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Cerium (IV) fluoride
58-0500-10G
58-0500-50G
100201-708EA
99.08
USD
100201-708
100207-418
Cerium tetrafluoride, anhydrous ≥99%
Cerium tetrafluoride
Formula:
CeF₄ MW: 216.11 Melting Pt: 650 (dec.) Density: 4.77 g/cm³ (20 °C) |
MDL Number:
MFCD00049422 CAS Number: 10060-10-3 Merck Index: 12,02034 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...