Lanthanum(III) nitrate hexahydrate ≥99.999% (La basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
57-5732-100G
57-5732-25G
100197-568EA
257.6
USD
100197-568
100206-772
Lanthanum(III) nitrate hexahydrate ≥99.999% (La basis), PURATREM
Lanthanum(III) nitrate hexahydrate
Formula:
La(NO₃)₃·6H₂O MW: 433.01 g/mol Boiling Pt: 126 (dec.) Melting Pt: 40 °C Density: 2.347 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149751 CAS Number: 10277-43-7 EINECS: 233-238-0 UN: 1477 ADR: 5.1 II Merck Index: 14,05363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...