Lanthanum(III) chloride hydrate ≥99.999% (La basis), PURATREM (low Ca, Fe, Mg)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Lanthanum trichloride hydrate

57-5730-25G 57-5730-100G
100206-770EA 135.88 USD
100206-770 100206-120
Lanthanum(III) chloride hydrate ≥99.999% (La basis), PURATREM (low Ca, Fe, Mg)
Lanthanum(III) chloride hydrate
Formula: Cl₃La·H₂O
MW: 263.28 g/mol
Boiling Pt: ∼860 °C (1013 hPa)
Melting Pt: 91 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149753
CAS Number: 20211-76-1
EINECS: 233-237-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR