Lanthanum(III) fluoride, anhydrous ≥99.99% (La basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Lanthanum trifluoride
57-2000-100G
57-2000-25G
100210-022EA
481.23
USD
100210-022
100210-024
Lanthanum(III) fluoride, anhydrous ≥99.99% (La basis), PURATREM
Lanthanum(III) fluoride
Formula:
F₃La MW: 195.9 g/mol Boiling Pt: 2327 Melting Pt: 1493 °C Density: 5.936 Storage Temperature: Ambient |
MDL Number:
MFCD00011073 CAS Number: 13709-38-1 EINECS: 237-252-8 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...