Barium carbonate ≥99.999% (Ba basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

56-1025-10G 56-1025-50G
100207-710EA 90.58 USD
100207-710 100199-966
Barium carbonate ≥99.999% (Ba basis), PURATREM
Barium carbonate
Formula: BaCO₃
Melting Pt: 1350 °C
Density: 3.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 13,00972

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR