Barium ≥99.7% (Sr <1.0%), pieces

Supplier: STREM CHEMICALS INC MS
Danger

56-0075-10G 56-0075-250G 56-0075-50G
100206-762EA 75.02 USD
100206-762 100206-764 100206-766
Barium ≥99.7% (Sr <1.0%), pieces
Barium
Formula: Ba
Boiling Pt: 1640 °C
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR