Barium ≥99.9%, dendritic pieces

Supplier: STREM CHEMICALS INC MS
Danger

56-0074-25G 56-0074-5G
100199-962EA 3065.72 USD
100199-962 100199-964
Barium ≥99.9%, dendritic pieces
Barium
Formula: Ba
Boiling Pt: 1640 °C
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR