Cesium ≥99.9%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Prescored ampoule.
55-5551-1G
55-5551-5G
55-5551-25G
100209-370EA
242.03
USD
100209-370
100210-654
100206-114
Cesium ≥99.9%
Cesium
Formula:
Cs MW: 132.91 Boiling Pt: 705 Melting Pt: 28.5 Density: 1.892 |
MDL Number:
MFCD00134037 CAS Number: 7440-46-2 EINECS: 231-155-4 UN: 1407 ADR: 4.3 I Merck Index: 12,02051 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...