Xenon difluoride ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Xenon(II) fluoride
54-1500-10G
54-1500-2G
100207-706EA
786.95
USD
100207-706
100207-708
Xenon difluoride ≥99.5%
Xenon difluoride
Halides for CVD & ALD, Volatile Precursors for CVD
Formula:
F₂Xe MW: 169.29 g/mol Boiling Pt: 114 °C (1013 hPa) Melting Pt: 128…130 °C Density: 4.32 Storage Temperature: Ambient |
MDL Number:
MFCD00040538 CAS Number: 13709-36-9 EINECS: 237-251-2 UN: 3085 ADR: 5.1 III Merck Index: 14,10072 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...