Triisopropyl borate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tri-i-propylborate

05-1400-100G 05-1400-500G
100206-202EA 53.78 USD
100206-202 100195-948
Triisopropyl borate ≥98%
Triisopropyl borate
Formula: C₉H₂₁BO₃
MW: 188.07 g/mol
Boiling Pt: 139…141 °C
Melting Pt: –59 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 50 °F
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS Number: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR