Triphenylantimony ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Triphenylstibine

51-3000-50G 51-3000-10G
100201-700EA 140.12 USD
100201-700 100210-004
Triphenylantimony ≥97%
Triphenylantimony
Formula: C₁₈H₁₅Sb
MW: 353.08 g/mol
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 52 - 54 °C
Density: 1.5 g/cm³ (12 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003000
CAS Number: 603-36-1
EINECS: 210-037-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR