3-Tolylboronic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

05-1049-25G 05-1049-5G
100199-704EA 873.29 USD
100199-704 100204-474
3-Tolylboronic acid ≥97%
3-Tolylboronic acid

Boronic Acid and Ester

Formula: C₇H₉BO₂
Melting Pt: 160…162 °C
MDL Number: MFCD00040198
CAS Number: 17933-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR