2-Thiopheneboronic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Thiophen-2-ylboronic acid, Thiophene-2-boronic acid, 2-Thienylboronic acid

05-1040-5G 05-1040-25G
100197-296EA 127.38 USD
100197-296 100197-294
2-Thiopheneboronic acid ≥97%
2-Thiopheneboronic acid

Boronic Acid and Ester

Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD00151850
CAS Number: 6165-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR