1-Naphthyl boronic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

05-1030-1G 05-1030-5G
100210-774EA 69.35 USD
100210-774 100199-698
1-Naphthyl boronic acid ≥97%
1-Naphthyl boronic acid

Boronic Acid and Ester

Formula: C₁₀H₉BO₂
Melting Pt: 210…211 °C
MDL Number: MFCD00019722
CAS Number: 13922-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR