Antimony ≥99.999%, shot

Supplier: STREM CHEMICALS INC MS
Warning

51-0090-50G 51-0090-10G 51-0090-250G
100206-758EA 212.31 USD
100206-758 100197-558 100195-916
Antimony ≥99.999%, shot
Antimony
Formula: Sb
Boiling Pt: 1380 °C
Melting Pt: 630 °C
Density: 6.684 g/cm³ (20 °C)
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III
Merck Index: 12,00733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR