Antimony ≥99.8%, rods

Supplier: STREM CHEMICALS INC MS
Warning

51-0085-1ROD 51-0085-5ROD
100206-756EA 178.34 USD
100206-756 100202-672
Antimony ≥99.8%, rods
Antimony
Formula: Sb
Boiling Pt: 1637 °C (1013 hPa)
Melting Pt: 630 °C
Density: 6.684 g/cm³ (20 °C)
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III
Merck Index: 12,00733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR