4-Methoxyphenylboronic acid ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
05-0880-25G
05-0880-5G
100203-694EA
587.38
USD
100203-694
100199-308
4-Methoxyphenylboronic acid ≥97%
4-Methoxyphenylboronic acid
Boronic Acid and Ester
Formula:
C₇H₉BO₃ Melting Pt: 204…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039139 CAS Number: 5720-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...