2-Methoxyphenylboronic acid ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
2-Methoxybenzeneboronic acid, (2-Methoxyphenyl)metaboric acid, Dihydroxy(2-methoxyphenyl)borane
05-0878-25G
05-0878-5G
100202-834EA
765.72
USD
100202-834
100209-288
2-Methoxyphenylboronic acid ≥97%
2-Methoxyphenylboronic acid
Boronic Acid and Ester
Formula:
C₇H₉BO₃ MW: 151.96 g/mol Melting Pt: 165 |
MDL Number:
MFCD00236047 CAS Number: 5720-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...