4-Fluorophenylboronic acid ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
05-0800-5G
05-0800-25G
100199-696EA
117.48
USD
100199-696
100199-694
4-Fluorophenylboronic acid ≥97%
4-Fluorophenylboronic acid
Boronic Acid and Ester
Formula:
C₆H₆BFO₂ Melting Pt: 263…265 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039136 CAS Number: 1765-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...