4-Chlorophenylboronic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4-Chlorobenzeneboronic acid, Dihydroxy(4-chlorophenyl)borane, (4-Chlorophenyl)boronic acid, (4-Chlorophenyl)metaboric acid

05-0430-5G 05-0430-25G
100200-372EA 84.92 USD
100200-372 100202-346
4-Chlorophenylboronic acid ≥97%
4-Chlorophenylboronic acid

Boronic Acid and Ester

Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 284…289 °C
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR