Trimethyltin bromide ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bromotrimethylstannane

50-3500-2G 50-3500-10G
100205-174EA 117.48 USD
100205-174 100202-766
Trimethyltin bromide ≥97%
Trimethyltin bromide
Formula: C₃H₉BrSn
MW: 243.72 g/mol
Boiling Pt: 61 °C (13 mmHg)
Melting Pt: 27 °C
MDL Number: MFCD00000051
CAS Number: 1066-44-0
EINECS: 213-916-2
UN: 3146
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR