Triethyltin chloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

50-3008-5G 50-3008-25G
100208-032EA 144.37 USD
100208-032 100203-800
Triethyltin chloride ≥98%
Triethyltin chloride
Formula: C₆H₁₅ClSn
MW: 241.35
Boiling Pt: 206 °C
Melting Pt: 15.5 °C
Density: 1.429
MDL Number: MFCD00040414
CAS Number: 994-31-0
EINECS: 213-616-1
UN: 2788
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR