Tetraisopropyltin 98

Supplier: STREM CHEMICALS INC MS

50-1950-10G 50-1950-2G
100207-408EA 635.51 USD
100207-408 100199-422
Tetraisopropyltin 98
Tetraisopropyltin
Formula: C₁₂H₂₈Sn
MW: 291.06
MDL Number: MFCD00026346
CAS Number: 2949-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR