Dimethyltin dichloride ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Dichlorodimethylstannane

50-1160-5G 50-1160-25G
100203-220EA 75.02 USD
100203-220 100199-958
Dimethyltin dichloride ≥95%
Dimethyltin dichloride
Formula: C₂H₆Cl₂Sn
MW: 219.69 g/mol
Boiling Pt: 188…190 °C
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00000517
CAS Number: 753-73-1
EINECS: 212-039-2
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR