Di-tert-butyltin dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Di-t-butyltin dichloride, Dichlorodi-tert-butylstannane

50-1140-1G 50-1140-5G
100196-306EA 58.03 USD
100196-306 100198-216
Di-tert-butyltin dichloride ≥98%
Di-tert-butyltin dichloride
Formula: C₈H₁₈Cl₂Sn
MW: 303.85 g/mol
Boiling Pt: 66 °C (3 mmHg)
Melting Pt: 42…43 °C
MDL Number: MFCD00009854
CAS Number: 19429-30-2
EINECS: 243-051-6
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR