Borane dimethyl sulfide complex contains 5-6% excess dimethylsulfide

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Borane-methyl sulphide complex

05-0110-100G 05-0110-25G
100207-924EA 184 USD
100207-924 100208-750
Borane dimethyl sulfide complex contains 5-6% excess dimethylsulfide
Borane dimethyl sulphide complex
Formula: C₂H₉BS
MW: 75.97 g/mol
Melting Pt: –40 °C
Density: 1.28 g/cm³ (20 °C)
Flash Pt: 65 °F
MDL Number: MFCD00013189
CAS Number: 13292-87-0
EINECS: 236-313-6
UN: 3148
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR