Indium ≥99.99%, foil
Supplier: STREM CHEMICALS INC MS
|
Warning
|
49-0050-100X100MM
49-0050-50X50MM
49-0050-150X300MM
100196-300EA
772.8
USD
100196-300
100208-030
100207-700
Indium ≥99.99%, foil
Indium
Formula:
In MW: 114.82 g/mol Boiling Pt: 2080 °C Melting Pt: 156.6 °C Density: 7.3 g/cm³ (20 °C) |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...