Cadmium oxide ≥99.999% (Cd basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cadmium (II) oxide
48-0800-500G
48-0800-100G
48-0800-25G
100195-912EA
1104
USD
100195-912
100206-740
100201-690
Cadmium oxide ≥99.999% (Cd basis), PURATREM
Cadmium oxide
Formula:
CdO MW: 128.41 g/mol Boiling Pt: 1559 °C (1013 hPa) Melting Pt: ~ 1426 °C Density: 8.15 g/cm³ (20 °C) |
MDL Number:
MFCD00010921 CAS Number: 1306-19-0 EINECS: 215-146-2 UN: 2570 ADR: 6.1,I Merck Index: 13,01623 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...