Silver ≥99.9%, wire

Supplier: STREM CHEMICALS INC MS

47-0350-25MT 47-0350-100MT
100197-548EA 144.37 USD
100197-548 100207-258
Silver ≥99.9%, wire
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.49
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR