Silver ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS

47-0055-200X200MM 47-0055-100X100MM 47-0055-50X50MM
100200-280EA 438.77 USD
100200-280 100197-542 100206-728
Silver ≥99.9%, foil
Silver
Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960.5 °C
Density: 10.49
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR