Silver ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS

47-0050-100X100MM 47-0050-25X25MM 47-0050-50X50MM
100206-110EA 1285.17 USD
100206-110 100201-688 100204-074
Silver ≥99.9%, foil
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.49
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR