Dipotassium tetrachloropalladate ≥99%

Supplier: STREM CHEMICALS INC MS

46-2126-5G 46-2126-1G
100201-682EA 462.83 USD
100201-682 100201-680
Dipotassium tetrachloropalladate ≥99%
Dipotassium tetrachloropalladate
Formula: Cl₄K₂Pd
Melting Pt: Dec.
Density: 2.67 g/cm³ (20 °C)
MDL Number: MFCD00011373
CAS Number: 10025-98-6
EINECS: 233-049-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR