Palladium ≥99.9%, shot - 6.35 mm
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-2116-5G
46-2116-1G
100202-406EA
2342.46
USD
100202-406
100205-158
Palladium ≥99.9%, shot - 6.35 mm
Palladium
Formula:
Pd Boiling Pt: 3167 °C (1013 hPa) Melting Pt: 1552 °C Density: 12.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 Merck Index: 12,07121 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...