Palladium 5% on barium sulfate, reduced

Supplier: STREM CHEMICALS INC MS

46-1989-50G 46-1989-10G
100199-948EA 857.72 USD
100199-948 100207-698
Palladium 5% on barium sulfate, reduced
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR