Palladium, black ≥99.9%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
46-1830-5G
46-1830-1G
100200-698EA
1286.58
USD
100200-698
100206-716
Palladium, black ≥99.9%
Palladium
Formula:
Pd Boiling Pt: 3140 °C Melting Pt: 1552 °C Density: 12.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 UN: 3089 ADR: 4.1,II Merck Index: 12,07121 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...