Sodium hexachlororhodate (III) hydrate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Rhodium sodium chloride hydrate

45-2000-5G 45-2000-250MG 45-2000-1G
100200-984EA 1268.18 USD
100200-984 100204-066 100205-954
Sodium hexachlororhodate (III) hydrate
Sodium hexachlororhodate (III) hydrate
Formula: Cl₆Na₃Rh·xH2O
MDL Number: MFCD00149196
CAS Number: 14972-70-4
EINECS: 239-047-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR