Rhodium(III) chloride hydrate (38 - 41% Rh)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Rhodium trichloride hydrate
45-1880-1G
45-1880-5G
45-1880-250MG
100209-356EA
744.49
USD
100209-356
100199-936
100208-438
Rhodium(III) chloride hydrate (38 - 41% Rh)
Rhodium (III) chloride hydrate
Formula:
Cl₃Rh·H₂O MW: 227.28 g/mol Melting Pt: 100 °C (dec.) |
MDL Number:
MFCD00149839 CAS Number: 20765-98-4 EINECS: 233-165-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...