Rhodium(III) iodide ≥99.9% (Rh basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
45-1805-1G
45-1805-5G
100208-912EA
266.09
USD
100208-912
100198-204
Rhodium(III) iodide ≥99.9% (Rh basis)
Rhodium(III) iodide
Formula:
I₃Rh MW: 483.62 g/mol Boiling Pt: 100 °C (1013 hPa) Density: 6.4 g/cm³ (20 °C) |
MDL Number:
MFCD00016311 CAS Number: 15492-38-3 EINECS: 239-521-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...