Ruthenium(III) chloride hydrate ≥99.9% (Ru basis) (40-43% Ru)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
44-5880-5G
44-5880-25G
44-5880-1G
100201-660EA
263.26
USD
100201-660
100209-262
100206-664
Ruthenium(III) chloride hydrate ≥99.9% (Ru basis) (40-43% Ru)
Ruthenium(III) chloride hydrate
Formula:
Cl₃Ru·xH₂O Melting Pt: 100 °C (dec.) Density: 3.11 g/cm³ (20 °C) |
MDL Number:
MFCD00149844 CAS Number: 14898-67-0 EINECS: 233-167-5 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...