Ruthenium ≥99.99%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-4350-5G
44-4350-1G
100211-098EA
860.55
USD
100211-098
100211-096
Ruthenium ≥99.99%, powder
Ruthenium
Formula:
Ru MW: 101.07 Boiling Pt: 3900 °C Melting Pt: 2310 °C Density: 12.3 |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 EINECS: 231-127-1 UN: 3089 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...