Ruthenium ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Warning

44-4275-1G 44-4275-5G
100197-524EA 138.71 USD
100197-524 100208-906
Ruthenium ≥99.9%, powder
Ruthenium
Formula: Ru
MW: 101.07
Boiling Pt: 3900 °C
Melting Pt: 2310 °C
Density: 12.3
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR