Ruthenium, black ≥99.9%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-3000-500MG
44-3000-2G
44-3000-10G
100197-520EA
86.34
USD
100197-520
100204-058
100211-094
Ruthenium, black ≥99.9%
Ruthenium
Formula:
Ru MW: 101.07 Boiling Pt: 3900 °C Melting Pt: 2310 °C Density: 12.3 |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 EINECS: 231-127-1 UN: 3089 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...