Ruthenium(IV) oxide hydrate
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-1980-5G
44-1980-1G
100199-912EA
302.89
USD
100199-912
100203-156
Ruthenium(IV) oxide hydrate
Ruthenium(IV) oxide hydrate
Formula:
O₂Ru·H₂O MW: 151.08 g/mol Density: 6.97 g/cm³ (20 °C) |
MDL Number:
MFCD00149846 CAS Number: 32740-79-7 EINECS: 234-840-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...