Ruthenium(III) acetylacetonate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-1800-5G
44-1800-1G
100206-658EA
479.82
USD
100206-658
100206-656
Ruthenium(III) acetylacetonate ≥99%
Ruthenium(III) acetylacetonate
Formula:
C₁₅H₂₁O₆Ru Melting Pt: 230…235 °C |
MDL Number:
MFCD00000030 CAS Number: 14284-93-6 EINECS: 238-193-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...