Ammonium hexachlororuthenate(IV) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Diammonium hexachlororuthenate

44-0020-1G 44-0020-5G
100198-188EA 104.74 USD
100198-188 100203-146
Ammonium hexachlororuthenate(IV) ≥99%
Ammonium hexachlororuthenate(IV)
Formula: H₈Cl₆N₂Ru
MW: 349.86 g/mol
MDL Number: MFCD00015962
CAS Number: 18746-63-9
EINECS: 242-552-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR