Molybdenum(VI) oxide ≥99.999% (Mo basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Molybdenum trioxide
42-1476-50G
42-1476-10G
100203-144EA
314.22
USD
100203-144
100198-186
Molybdenum(VI) oxide ≥99.999% (Mo basis), PURATREM
Molybdenum(VI) oxide
Formula:
MoO₃ MW: 143.94 g/mol Boiling Pt: (subl.) Melting Pt: 795 °C Density: 4.69 g/cm³ (20 °C) |
MDL Number:
MFCD00003469 CAS Number: 1313-27-5 EINECS: 215-204-7 UN: 3288 ADR: 6.1,III Merck Index: 13,06261 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...