Molybdenum ≥99.95%, sheets

Supplier: STREM CHEMICALS INC MS
Warning

42-0080-50X50MM 42-0080-100X100MM 42-0080-500X100MM
100209-928EA 154.28 USD
100209-928 100201-628 100207-674
Molybdenum ≥99.95%, sheets
Molybdenum
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.2
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR