Molybdenum ≥99.95%, foil

Supplier: STREM CHEMICALS INC MS
Warning

42-0070-300X300MM 42-0070-150X300MM 42-0070-100X100MM
100209-926EA 469.91 USD
100209-926 100203-132 100205-118
Molybdenum ≥99.95%, foil
Molybdenum
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.2
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR