Niobium(IV) oxide ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Niobium dioxide

41-4110-10G 41-4110-2G 41-4110-50G
100200-460EA 205.23 USD
100200-460 100198-754 100208-428
Niobium(IV) oxide ≥99%
Niobium(IV) oxide
Formula: NbO₂
Melting Pt: 1915 °C
Density: 5.98 g/cm³ (20 °C)
MDL Number: MFCD00016242
CAS Number: 12034-59-2
EINECS: 234-809-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR